Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactose, Monohydrate, Spray-Dried Powder, NF, Spectrum™ Chemical
SDP

Catalog No. 18603701 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-701 25 kg Poly Pail
18-603-698 500 g Poly Pail
18-603-699 2.5 kg Poly Pail
18-603-700 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-701 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. LA10525KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LA105, 64044-51-5, C12H22O11·H2O

Spectrum™ Chemical Lactose, Monohydrate, Spray-Dried Powder, NF is used to help form tablets because it has excellent compressibility properties and as a diluent powder for dry-powder inhalations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
InChI Key WSVLPVUVIUVCRA-RJMJUYIDSA-N
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
SMILES O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Poly Pail
Quantity 25 kg
Specific Rotation 110.3
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Lactose, Monohydrate, Spray-Dried Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.