Learn More
HIMEDIA Lactose Monohydrate, Sterile, RM565G, Microbiology Culture Media Ingredients, Carbohydrate, Gamma Irradiated Sterile, CAS 64044-51-5, Fermentation Substrate, Powder, 500 g
Catalog No. NC4013787
Encompass_Preferred
Lactose monohydrate, Sterile is a gamma-irradiated, low-bioburden carbohydrate (CAS 64044-51-5, molecular weight 360.31) supplied as a pale yellow fine powder that gives a clear solution at 10% in boiling water. As a sterile, ready-to-use fermentable sugar it is ideal where added bioburden cannot be tolerated, serving as a carbon source and excipient in microbiological media and in pharmaceutical and biotechnology applications. Store below 30 C, shelf life two years. Packaged as 500 g.
Catalog No. NC4013787 Supplier HIMEDIA Supplier No. RM565G500G
Lactose monohydrate, Sterile is a gamma-irradiated, low-bioburden carbohydrate (CAS 64044-51-5, molecular weight 360.31) supplied as a pale yellow fine powder that gives a clear solution at 10% in boiling water. As a sterile, ready-to-use fermentable sugar it is ideal where added bioburden cannot be tolerated, serving as a carbon source and excipient in microbiological media and in pharmaceutical and biotechnology applications. Store below 30 C, shelf life two years. Packaged as 500 g.