Learn More
U.S. Pharmacopeia Lactose monohydrate, 64044-51-5, MFCD00166994, 500 mg
Supplier:  U.S. Pharmacopeia 1356701
Encompass
Empirical Formula (Hill Notation): C12H22O11 · H2O, Molecular Weight: 360.31, Synonym: β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, D-Lactose monohydrate, Milk sugar.
Catalog No. NC9882862