Learn More
Description
Lactose NF is an odorless, crystalline powder that is white or almost white.
Alternative name(s): Lactose Monohydrate
Specifications
Specifications
| Chemical Name or Material | Lactose NF (Monohydrate) |
| Packaging | Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck |
| CAS | 64044-51-5 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 360.3 |
| Quantity | 1 kg |
| Grade | NF |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
