Learn More
Medchemexpress LLC Lactose octaacetate | 6291-42-5 | 98.0% | 678.59 | 100 G
SDP
Catalog No. 5000452201
Encompass_Preferred
Lactose octaacetate exhibits mild to moderate antifungal activity against certain fungi, but demonstrates low or no activity against bacteria and yeast. It shows low cytotoxicity to MDBK cells, Hep-2, and MDCK cells, and possesses antiviral activity against PV-1.
  • Shows mild to moderate antifungal activity against some fungi
  • Has low or no activity against bacteria and yeast
  • Shows low cytotoxicity to MDBK cells, Hep-2 and MDCK cells
  • Has antiviral activity against PV-1
  • Appearance: solid, white to off-white
Catalog No. 50-004-52201 Supplier Medchemexpress LLC Supplier No. HYW008864100G
Lactose octaacetate exhibits mild to moderate antifungal activity against certain fungi, but demonstrates low or no activity against bacteria and yeast. It shows low cytotoxicity to MDBK cells, Hep-2, and MDCK cells, and possesses antiviral activity against PV-1.
  • Shows mild to moderate antifungal activity against some fungi
  • Has low or no activity against bacteria and yeast
  • Shows low cytotoxicity to MDBK cells, Hep-2 and MDCK cells
  • Has antiviral activity against PV-1
  • Appearance: solid, white to off-white