Learn More
Medchemexpress LLC Lactose octaacetate | 6291-42-5 | 98.0% | 678.59 | 25 G
SDP
Catalog No. 5000453291
Encompass_Preferred
Lactose octaacetate exhibits mild to moderate antifungal activity against certain fungi, but it shows low or no activity against bacteria and yeast. It also demonstrates low cytotoxicity to MDBK cells, Hep-2, and MDCK cells, and possesses antiviral activity against PV-1.
  • Mild to moderate antifungal activity against some fungi.
  • Low or no activity against bacteria and yeast.
  • Low cytotoxicity to MDBK cells, Hep-2, and MDCK cells.
  • Antiviral activity against PV-1.
Catalog No. 50-004-53291 Supplier Medchemexpress LLC Supplier No. 25GHYW008864
Lactose octaacetate exhibits mild to moderate antifungal activity against certain fungi, but it shows low or no activity against bacteria and yeast. It also demonstrates low cytotoxicity to MDBK cells, Hep-2, and MDCK cells, and possesses antiviral activity against PV-1.
  • Mild to moderate antifungal activity against some fungi.
  • Low or no activity against bacteria and yeast.
  • Low cytotoxicity to MDBK cells, Hep-2, and MDCK cells.
  • Antiviral activity against PV-1.