Learn More
Bioworld LACTOSE SEPAROPOREAGAROSE
Catalog No. 50197702
Encompass
LACTOSE SEPAROPORE (AGAROSE) 6B-CL: Lactose Coupled Separopore 6B-CL Bio-World's Lactose Separopore 6Bis prepared by epoxy activation of Separopore. Lactose-Separopore may be used to isolate lactose binding proteins, such as lectins.; Pl...
Catalog No. 50-197-702 Supplier Bioworld Supplier No. 201810172
LACTOSE SEPAROPORE (AGAROSE) 6B-CL: Lactose Coupled Separopore 6B-CL Bio-World's Lactose Separopore 6Bis prepared by epoxy activation of Separopore. Lactose-Separopore may be used to isolate lactose binding proteins, such as lectins.; Pl...