Learn More
Cayman Chemical LactosylcRamID nd GnglIos 1mL
SDP
Catalog No. 502990035
Encompass_Preferred
A mixture of lactosylceramides and sialogangliosides; contains LacCers, the monosialoganglioside GM3, and the disialoganglioside GD3; intended for use as a reference standard for the detection of LacCers, GM3, and GD3 in various sample types by TLC, LC-, or GC-MS
Catalog No. 50-299-0035 Supplier Cayman Chemical Supplier No. 293611 ML
A mixture of lactosylceramides and sialogangliosides; contains LacCers, the monosialoganglioside GM3, and the disialoganglioside GD3; intended for use as a reference standard for the detection of LacCers, GM3, and GD3 in various sample types by TLC, LC-, or GC-MS