Learn More
Cayman Chemical Lactoyl PhenylalnIn 10mg
SDP
Catalog No. 502918018
Encompass_Preferred
A secondary metabolite; decreases food intake, body weight, and adiposity, as well as improves glucose homeostasis in a mouse model of high-fat diet-induced obesity at 50 mg/kg per day; plasma levels of lactoyl phenylalanine are increased following exercise in mice
Catalog No. 50-291-8018 Supplier Cayman Chemical Supplier No. 3730410 MG
A secondary metabolite; decreases food intake, body weight, and adiposity, as well as improves glucose homeostasis in a mouse model of high-fat diet-induced obesity at 50 mg/kg per day; plasma levels of lactoyl phenylalanine are increased following exercise in mice