Learn More
Cayman Chemical Lactoyl Phenylalanine, Purity: ≥95%, CAS: 183241-73-8, 50 mg
SDP
Catalog No. 502917449
Encompass_Preferred
A secondary metabolite; decreases food intake, body weight, and adiposity, as well as improves glucose homeostasis in a mouse model of high-fat diet-induced obesity at 50 mg/kg per day; plasma levels of lactoyl phenylalanine are increased following exercise in mice
Catalog No. 50-291-7449 Supplier Cayman Chemical Supplier No. 3730450 MG
A secondary metabolite; decreases food intake, body weight, and adiposity, as well as improves glucose homeostasis in a mouse model of high-fat diet-induced obesity at 50 mg/kg per day; plasma levels of lactoyl phenylalanine are increased following exercise in mice