Learn More
Medchemexpress LLC Lactyl-CoA | 1926-57-4 | 96.92% | C24H40N7O18P3S | 1 ML
SDP
Catalog No. 5000353413
Encompass_Preferred
Lactyl-CoA is an acyl-CoA essential for the biosynthesis of biodegradable and biocompatible lactic acid-based copolymers. It is formally condensed from the sulfhydryl group of CoA and the carboxyl group of lactic acid. In vitro, GTPSCS can be used as Lactyl-CoA synthetase, converting lactic acid into Lactyl-CoA and providing substrate for histone lactation.
  • Essential for biosynthesis of biodegradable and biocompatible lactic acid-based copolymers.
  • Acts as a substrate for histone lactation.
  • Store at -80°C, protected from light, and under nitrogen.
Catalog No. 50-003-53413 Supplier Medchemexpress LLC Supplier No. HY14154010MM/1ML
Lactyl-CoA is an acyl-CoA essential for the biosynthesis of biodegradable and biocompatible lactic acid-based copolymers. It is formally condensed from the sulfhydryl group of CoA and the carboxyl group of lactic acid. In vitro, GTPSCS can be used as Lactyl-CoA synthetase, converting lactic acid into Lactyl-CoA and providing substrate for histone lactation.
  • Essential for biosynthesis of biodegradable and biocompatible lactic acid-based copolymers.
  • Acts as a substrate for histone lactation.
  • Store at -80°C, protected from light, and under nitrogen.