Learn More
Medchemexpress LLC Lactyl-CoA | 1926-57-4 | 98.0% | C24H40N7O18P3S | 50 MG
SDP
Catalog No. 5000353765
Encompass_Preferred
Lactyl-CoA is an acyl-CoA formed by the condensation of the sulfhydryl group of CoA and the carboxyl group of lactic acid. It is essential for the biosynthesis of biodegradable and biocompatible lactic acid-based copolymers.
  • Essential for the biosynthesis of lactic acid-based copolymers
  • Formed from CoA and lactic acid
Catalog No. 50-003-53765 Supplier Medchemexpress LLC Supplier No. HY14154050MG
Lactyl-CoA is an acyl-CoA formed by the condensation of the sulfhydryl group of CoA and the carboxyl group of lactic acid. It is essential for the biosynthesis of biodegradable and biocompatible lactic acid-based copolymers.
  • Essential for the biosynthesis of lactic acid-based copolymers
  • Formed from CoA and lactic acid