Learn More
Medchemexpress LLC Lactyl-CoA | 1926-57-4 | 98.01% | C24H40N7O18P3S | 25 MG
SDP
Catalog No. 5000352532
Encompass_Preferred
Lactyl-CoA is an acyl-CoA formed by the condensation of the sulfhydryl group of CoA and the carboxyl group of lactic acid. It is crucial for the biosynthesis of biodegradable and biocompatible lactic acid-based copolymers. GTPSCS can also function as Lactyl-CoA synthetase, converting lactic acid into Lactyl-CoA to provide substrate for histone lactation.
Catalog No. 50-003-52532 Supplier Medchemexpress LLC Supplier No. 25MGHY141540
Lactyl-CoA is an acyl-CoA formed by the condensation of the sulfhydryl group of CoA and the carboxyl group of lactic acid. It is crucial for the biosynthesis of biodegradable and biocompatible lactic acid-based copolymers. GTPSCS can also function as Lactyl-CoA synthetase, converting lactic acid into Lactyl-CoA to provide substrate for histone lactation.