Learn More
Electron Microscopy Sciences Lacey Carbon Film with Carbon/Formvar 1 nm / 5 nm thick on 400 mesh Copper Grid
SDP
Catalog No. 501931358
Encompass_Preferred
The continuous ultrathin carbon/formvar film on lacey carbon allows for the thinnest support film that still has adequate strength to withstand electron microscope specimen preparation. The carbon 1 nm, formvar 5 nm film lies across a carbon lacey film supported by a mesh grid. The size of the holes in the lacey film differ widely from batch to batch but are generally in the range of ¼ µm to 5µm, which gives the equivalent support of at least 6000 mesh grid.
Specimen material lying over the covered holes can be imaged in the TEM with practically no interference from the carbon film supporting it. This product is ideal for looking at nanotubes, virus particles and other small particulate material.
Grid: Copper
Catalog No. 50-193-1358 Supplier Electron Microscopy Sciences Supplier No. LC400CUFCF
The continuous ultrathin carbon/formvar film on lacey carbon allows for the thinnest support film that still has adequate strength to withstand electron microscope specimen preparation. The carbon 1 nm, formvar 5 nm film lies across a carbon lacey film supported by a mesh grid. The size of the holes in the lacey film differ widely from batch to batch but are generally in the range of ¼ µm to 5µm, which gives the equivalent support of at least 6000 mesh grid.
Specimen material lying over the covered holes can be imaged in the TEM with practically no interference from the carbon film supporting it. This product is ideal for looking at nanotubes, virus particles and other small particulate material.
Grid: Copper