Learn More
Selleck Chemical LLC Ladarixin E4650-1G
SDP
Catalog No. 5000038978
Encompass_Preferred
Ladarixin(DF 2156A free base) is a small molecule orally available allosteric and non-competitive antagonist of dual CXCR1 and CXCR2 It is able to reduce the acute and chronic neutrophilic influx and can be used in research of Asthma Idiopathic Pulmonary Fibrosis Influenza-A infection and COPD cutaneous and uveal melanoma conditions
Catalog No. 50-000-38978 Supplier Selleck Chemical LLC Supplier No. E46501G
Ladarixin(DF 2156A free base) is a small molecule orally available allosteric and non-competitive antagonist of dual CXCR1 and CXCR2 It is able to reduce the acute and chronic neutrophilic influx and can be used in research of Asthma Idiopathic Pulmonary Fibrosis Influenza-A infection and COPD cutaneous and uveal melanoma conditions