Learn More
Grainger GEMTOR Ladder Escape Belt, Nylon, Quick Connect, Universal, 1 ea
Catalog No. 502795447
Encompass_Preferred
This ladder and escape belt is designed for use as a positioning device while on a ladder, as an emergency self-rescue device, or as an anchor tie-off. It features high-strength construction to ensure reliability in critical rescue and positioning operations.
  • Constructed from 1.75 in. wide nylon webbing with a 3 in. wide durable nylon web back pad
  • Lock stitched with high tenacity nylon thread for enhanced durability
  • Adjustable for waist sizes from 32 in. to 54 in.
  • Equipped with an aluminum 3-stage locking carabiner with a 2 in. side opening gate
  • UL Certified to NFPA 2500/1983-2022 edition
  • Includes aluminum pompier hook
Catalog No. 50-279-5447 Supplier Grainger Supplier No. 19TJ12
This ladder and escape belt is designed for use as a positioning device while on a ladder, as an emergency self-rescue device, or as an anchor tie-off. It features high-strength construction to ensure reliability in critical rescue and positioning operations.
  • Constructed from 1.75 in. wide nylon webbing with a 3 in. wide durable nylon web back pad
  • Lock stitched with high tenacity nylon thread for enhanced durability
  • Adjustable for waist sizes from 32 in. to 54 in.
  • Equipped with an aluminum 3-stage locking carabiner with a 2 in. side opening gate
  • UL Certified to NFPA 2500/1983-2022 edition
  • Includes aluminum pompier hook