Learn More
Grainger Gemtor Ladder Escape Belt, Nylon, Tongue Buckle, Size XS, 1 ea
Catalog No. 502796179
Encompass_Preferred
This ladder rescue escape belt is designed for secure positioning and self-anchoring during firefighting and rescue operations. Featuring a robust nylon construction and a steel tongue-buckle design, this belt provides reliable performance for emergency responders.
  • Fits waist sizes 34 in to 40 in
  • Includes a zinc plated steel triple action pompier hook with a 1 in. gate opening
  • UL classified and meets NFPA 1983-2012 edition standards
  • Hardware features 50 hour salt spray corrosion resistance
  • Belt width of 1 3/4 in. and body pad width of 5 in.
  • Single anchor point configuration
Catalog No. 50-279-6179 Supplier Grainger Supplier No. 19TJ03
This ladder rescue escape belt is designed for secure positioning and self-anchoring during firefighting and rescue operations. Featuring a robust nylon construction and a steel tongue-buckle design, this belt provides reliable performance for emergency responders.
  • Fits waist sizes 34 in to 40 in
  • Includes a zinc plated steel triple action pompier hook with a 1 in. gate opening
  • UL classified and meets NFPA 1983-2012 edition standards
  • Hardware features 50 hour salt spray corrosion resistance
  • Belt width of 1 3/4 in. and body pad width of 5 in.
  • Single anchor point configuration