Learn More
BURKLE INC Detectable ladle, long handle, 1.01 oz. (30 ml), PS, sterile
Catalog No. 503081362
Encompass_Preferred
In addition to the blue color, these samplers have a special additive in the material that makes them visible in metal detectors or by X-ray. Although they are made of plastic, these products can therefore be sorted out using the usual systems for checking for foreign bodies.
The scoop is suitable for sampling liquids and viscous media as well as powders and granulates. The ladle has a smooth bottom edge and rounded corners. This makes it possible to reach every nook and cranny in the container or to pick up medium directly from the bottom of the container.
A long handle with finger notches ensures a good grip and precise work.
Catalog No. 50-308-1362 Supplier BURKLE INC Supplier No. 53785024
In addition to the blue color, these samplers have a special additive in the material that makes them visible in metal detectors or by X-ray. Although they are made of plastic, these products can therefore be sorted out using the usual systems for checking for foreign bodies.
The scoop is suitable for sampling liquids and viscous media as well as powders and granulates. The ladle has a smooth bottom edge and rounded corners. This makes it possible to reach every nook and cranny in the container or to pick up medium directly from the bottom of the container.
A long handle with finger notches ensures a good grip and precise work.