Learn More
BURKLE INC Ladle, long handle, 1.01 oz. (30 ml), PS, white, sterile
Supplier:  BURKLE INC 53785022
Encompass_Preferred
The single-use ladle is perfect for sampling liquids, viscous media, powders, and granulates. Its smooth base and rounded corners allow complete emptying of containers, reaching every edge and the bottom with ease.
The long handle with ergonomic finger recesses ensures a secure grip, precise handling, and safe distance to avoid contamination or hazards.
The extra-long version enables sampling from deep containers or hard-to-reach areas.
Disposable design means no cleaning or sterilization, saving time, reducing costs, and improving safety by minimizing exposure to hazardous substances. Eliminating cleaning avoids handling sharp or dirty reusable tools.
Individually packaged, each ladle remains sterile and contamination-free until use, preventing cross-contamination between samples or products.
Catalog No. 50-308-1226