Learn More
Medchemexpress LLC Ladostigil hydrochlo 5mg | 209394-18-3 | 5MG
SDP
Catalog No. 5000244160
Encompass_Preferred
Ladostigil (TV-3326) hydrochloride is an orally active dual inhibitor of cholinesterase and brain-selective monoamine oxidase (MAO) with IC50s of 37 1 and 31 8 M for MAO-B and AChE respectively Ladostigil hydrochloride exhibits neuroprotective antioxidant and anti-inflammatory activities Ladostigil can be used for the research of depression and Alzheimer s disease[1 [2 Ladostigil (hydrochloride) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-44160 Supplier Medchemexpress LLC Supplier No. HY10399A5MG
Ladostigil (TV-3326) hydrochloride is an orally active dual inhibitor of cholinesterase and brain-selective monoamine oxidase (MAO) with IC50s of 37 1 and 31 8 M for MAO-B and AChE respectively Ladostigil hydrochloride exhibits neuroprotective antioxidant and anti-inflammatory activities Ladostigil can be used for the research of depression and Alzheimer s disease[1 [2 Ladostigil (hydrochloride) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups