Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ladostigil hydrochloride | 209394-18-3 | >98.0% | 308.80 g·mol⁻1 | C16H21ClN2O2 | 25 MG
SDP

Catalog No. 5000243782
Encompass_Preferred

Ladostigil hydrochloride is the hydrochloride salt of ladostigil (TV-3326), a multifunctional, orally active research compound that combines cholinesterase inhibition with brain-selective monoamine oxidase inhibition. Provided as a white to off-white powder for research use only, it is used in preclinical studies of neuroprotection, cognitive impairment, and depression-related models.

  • Dual activity: cholinesterase and monoamine oxidase inhibition.
  • Hydrochloride salt form for increased solubility and stability.
  • Purity >98.0%.
  • Molecular weight 308.80 g·mol⁻1.
  • White to off-white powder suitable for formulation and dosing studies.
  • Certificate of analysis typically available on request.

Catalog No. 50-002-43782 Supplier Medchemexpress LLC Supplier No. HY10399A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ladostigil hydrochloride is the hydrochloride salt of ladostigil (TV-3326), a multifunctional, orally active research compound that combines cholinesterase inhibition with brain-selective monoamine oxidase inhibition. Provided as a white to off-white powder for research use only, it is used in preclinical studies of neuroprotection, cognitive impairment, and depression-related models.

  • Dual activity: cholinesterase and monoamine oxidase inhibition.
  • Hydrochloride salt form for increased solubility and stability.
  • Purity >98.0%.
  • Molecular weight 308.80 g·mol⁻1.
  • White to off-white powder suitable for formulation and dosing studies.
  • Certificate of analysis typically available on request.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.