Learn More
Medchemexpress LLC Ladostigil hydrochloride | 209394-18-3 | 99.9% | 308.80 g·mol⁻¹ | C16H21ClN2O2 | 10 MG
SDP
Catalog No. 5000243865
Encompass_Preferred
Ladostigil hydrochloride is an orally active dual inhibitor of cholinesterase and brain-selective monoamine oxidase, exhibiting neuroprotective, antioxidant, and anti-inflammatory activities. It is used in research on depression and neurodegenerative disorders, and its hydrochloride salt includes an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition for click-chemistry applications.
  • Dual inhibitor of cholinesterase and monoamine oxidase.
  • Neuroprotective, antioxidant, and anti-inflammatory activities.
  • Suitable for research on depression and neurodegeneration.
  • Contains an alkyne group for click-chemistry conjugation.
  • Provided with analytical documentation such as data sheet and COA.
Catalog No. 50-002-43865 Supplier Medchemexpress LLC Supplier No. HY10399A10MG
Ladostigil hydrochloride is an orally active dual inhibitor of cholinesterase and brain-selective monoamine oxidase, exhibiting neuroprotective, antioxidant, and anti-inflammatory activities. It is used in research on depression and neurodegenerative disorders, and its hydrochloride salt includes an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition for click-chemistry applications.
  • Dual inhibitor of cholinesterase and monoamine oxidase.
  • Neuroprotective, antioxidant, and anti-inflammatory activities.
  • Suitable for research on depression and neurodegeneration.
  • Contains an alkyne group for click-chemistry conjugation.
  • Provided with analytical documentation such as data sheet and COA.