Learn More
Medchemexpress LLC Ladostigil hydrochloride | 209394-18-3 | 99.9% | 308.80 g/mol | C16H21ClN2O2 | 50 MG
SDP
Catalog No. 5000243566
Encompass_Preferred
Ladostigil hydrochloride is the hydrochloride salt of ladostigil, an orally active dual inhibitor of cholinesterase and brain-selective monoamine oxidase. It exhibits neuroprotective, antioxidant, and anti-inflammatory properties and is used as a research reagent in studies of neurodegeneration, depression, and Alzheimer's disease.
  • Dual cholinesterase and monoamine oxidase inhibitor
  • Neuroprotective, antioxidant, and anti-inflammatory activity
  • Hydrochloride salt for improved solubility
  • Click-chemistry compatible (contains an alkyne group)
  • High purity verified by RP-HPLC, suitable for research
Catalog No. 50-002-43566 Supplier Medchemexpress LLC Supplier No. HY10399A50MG
Ladostigil hydrochloride is the hydrochloride salt of ladostigil, an orally active dual inhibitor of cholinesterase and brain-selective monoamine oxidase. It exhibits neuroprotective, antioxidant, and anti-inflammatory properties and is used as a research reagent in studies of neurodegeneration, depression, and Alzheimer's disease.
  • Dual cholinesterase and monoamine oxidase inhibitor
  • Neuroprotective, antioxidant, and anti-inflammatory activity
  • Hydrochloride salt for improved solubility
  • Click-chemistry compatible (contains an alkyne group)
  • High purity verified by RP-HPLC, suitable for research