Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Laduviglusib (CHIR-99021) | 252917-06-9 | MFCD11846251 | 99.1% | 465.3 g·mol⁻¹ | C22H18Cl2N8 | 5 MG
SDP

Catalog No. 5000249264
Encompass_Preferred

Laduviglusib (CHIR-99021) analytical standard is a potent, selective GSK-3α/β inhibitor supplied as a solid reference material for research and analytical applications. It is intended for use as a reference standard in assay development, compound characterization, and analytical calibration, and is provided in small pack sizes suitable for laboratory workflows.

  • Analytical standard for assay development and compound characterization
  • High purity, 99.1% (assay) for reliable analytical performance
  • Off-white to light yellow solid suitable for weighing and formulation
  • Small pack size available (5 MG) for reference use
  • Molecular formula C22H18Cl2N8; molecular weight 465.3 g·mol⁻¹

Catalog No. 50-002-49264 Supplier Medchemexpress LLC Supplier No. HY10182R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laduviglusib (CHIR-99021) analytical standard is a potent, selective GSK-3α/β inhibitor supplied as a solid reference material for research and analytical applications. It is intended for use as a reference standard in assay development, compound characterization, and analytical calibration, and is provided in small pack sizes suitable for laboratory workflows.

  • Analytical standard for assay development and compound characterization
  • High purity, 99.1% (assay) for reliable analytical performance
  • Off-white to light yellow solid suitable for weighing and formulation
  • Small pack size available (5 MG) for reference use
  • Molecular formula C22H18Cl2N8; molecular weight 465.3 g·mol⁻¹

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.