Learn More
Medchemexpress LLC HY-10182A 50mg , Laduviglusib (monohydrochloride) CAS:1797989-42-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10182A50MG
Encompass_Preferred
HY-10182A 50mg Laduviglusib (monohydrochloride) CAS: 1797989-42-4 Laduviglusib (CHIR-99021) monohydrochloride is a potent and selective GSK-3α/β inhibitor with IC50s of 10 nM and 6.7 nM. Laduviglusib monohydrochloride shows >500-fold selectivity for GSK-3 over CDC2, ERK2 and other protein kinases. Laduviglusib monohydrochloride is also a potent Wnt/β-catenin signaling pathway activator. Laduviglusib monohydrochloride enhances mouse and human embryonic stem cells self-renewal. Laduviglusib monohydrochloride induces autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0415