Learn More
TARGETMOL CHEMICALS INC LADUVIGLUSIB TRIHYDROCHL 5MG
SDP
Catalog No. 502580937
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Laduviglusib trihydrochloride (CT99021 trihydrochloride) is a selective glycogen synthase kinase 3 (GSK-3) inhibitor with IC50s of 10 nM and 6.7 nM for GSK-3(alpha) and GSK-3(beta). Laduviglusib trihydrochloride inhibits human GSK-3(beta) with a Ki of 9.8 nM. Laduviglusib trihydrochloride is an activator of Wnt/(beta)-catenin signaling. Laduviglusib trihydrochloride induces autophagy. purity: 99%
Catalog No. 50-258-0937 Supplier TARGETMOL CHEMICALS INC Supplier No. T226575MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Laduviglusib trihydrochloride (CT99021 trihydrochloride) is a selective glycogen synthase kinase 3 (GSK-3) inhibitor with IC50s of 10 nM and 6.7 nM for GSK-3(alpha) and GSK-3(beta). Laduviglusib trihydrochloride inhibits human GSK-3(beta) with a Ki of 9.8 nM. Laduviglusib trihydrochloride is an activator of Wnt/(beta)-catenin signaling. Laduviglusib trihydrochloride induces autophagy. purity: 99%