Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ladybug Land
SDPSCI_ED

Catalog No. S79118 Shop All Insect Lore Products Products
Change view
Click to view available options
L x W x H: 10 x 7.5 x 8 in. (25 x 18.8 x 20cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S79118 L x W x H: 10 x 7.5 x 8 in. (25 x 18.8 x 20cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S79118 Supplier Insect Lore Products Supplier No. 2100
This item is not returnable. View return policy

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

  • Sturdy plastic habitat base
  • See-through magnifier plastic dome cap
  • Instructions
  • Mail-in certificate for 15 to 20 pink-spotted ladybug larvae and special food

Discover the land of the ladybugs.

  • Watch ladybugs grow from larvae to adults
  • Dimensions (L x W x H): 10 x 7.5 x 8 in. (25 x 18.8 x 20cm)

Order Info

Grades PreK+. Allow two weeks for delivery. Available in continental U.S. and Alaska only. Redemption fee applies when certificate is mailed in. Postage and handling fee required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.