Learn More
Crescent Chemical Co Inc LAEMMLI BUFFER 10X FOR SDS-PG
SDP
Supplier:  Crescent Chemical Co Inc SE42556.04
Encompass_Preferred
Laemmli Buffer 10X, for SDS-PAGE Running buffer for SDS PAGE. Supplied as 10 x concentrate. Contains 0.25 M Tris, 1.92 M glycine and 1 % SDS in aqueous solution. Size - 10L Storage Conditions - +15 °C TO +30 °C Catalog Number - 42556.04
Catalog No. 50-248-612