Learn More
Crescent Chemical Co Inc LAEMMLI SAMPLE BUFFER (2X)
SDP
Supplier:  Crescent Chemical Co Inc SE42526.02
Encompass_Preferred
Laemmli Sample Buffer (2x), for SDS PAGE Sample buffer for SDS PAGE. Supplied as 2x concentrate. Contains 126 mM Tris/Cl (pH 6.8), 20 % glycerol, 4 % SDS and 0.02 % bromophenol blue. Size - 5x20ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42526.02
Catalog No. 50-248-590