Learn More
Bioworld Laemmli SDS-Sample Buffer (4X), Reducing, 50 mL
Catalog No. 50196783
Encompass_Preferred
Laemmli (SDS-Sample) 4X Buffer, Reducing An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Technical Information:Ultra Pure GradeContents of a 1X Solution: 1.5% SDS and 10% glycerol, 62.5mM Tris -HCl (pH6.8), 0.0025% Bromophenol blue, 2% β-mercaptoethanol.DNase: None detectedRNase: None detectedProteases: None detected
Catalog No. 50-196-783 Supplier Bioworld Supplier No. 105700202
Laemmli (SDS-Sample) 4X Buffer, Reducing An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Technical Information:Ultra Pure GradeContents of a 1X Solution: 1.5% SDS and 10% glycerol, 62.5mM Tris -HCl (pH6.8), 0.0025% Bromophenol blue, 2% β-mercaptoethanol.DNase: None detectedRNase: None detectedProteases: None detected