Learn More
Bioworld Laemmli SDS-Sample Buffer (6X), Reducing, 50 mL
Catalog No. 50196785
Encompass_Preferred
Laemmli (SDS-Sample) 6X Buffer, Reducing An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Technical Information:Ultra Pure GradeContents of a 1X Solution: 1.5% SDS and 10% glycerol, 62.5mM Tris -HCl (pH 6.8), 0.0025% Bromophenol blue, 2% β-mercaptoethanol.DNase: None detectedRNase: None detectedProteases: None detectedStability (-20°C) >1 Year
Catalog No. 50-196-785 Supplier Bioworld Supplier No. 105700212
Laemmli (SDS-Sample) 6X Buffer, Reducing An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Technical Information:Ultra Pure GradeContents of a 1X Solution: 1.5% SDS and 10% glycerol, 62.5mM Tris -HCl (pH 6.8), 0.0025% Bromophenol blue, 2% β-mercaptoethanol.DNase: None detectedRNase: None detectedProteases: None detectedStability (-20°C) >1 Year