Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Laemmli SDS Sample Buffer, 4X

Catalog No. 04822252 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-822-252 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-822-252 Supplier MP Biomedicals™ Supplier No. 04822252
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laemmli SDS Sample Buffer, 4X is a sample loading buffer for protein gel electrophoresis.

  • Premixed liquid 4X concentrate.
  • Contains 250 mM TRIS-HCl (pH 6.8), 8% SDS, 40% glycerol, and 0.02% bromophenol blue.
  • For protein sample preparation in the Laemmli SDS-PAGE system, without reducing the disulfide linkages.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.