Learn More
Bioworld Laemmli SDS-Sample Buffer (4X), Non-Reducing, 25 mL
Catalog No. 50196778
Encompass_Preferred
Laemmli (SDS-Sample) 4X Buffer, Non-Reducing An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Composition of this buffer is similar to the reducing buffer minus mercaptoethanol. Technical Information:Ultra Pure GradeContents of a 1X Solution: 2% SDS and 10% glycerol, 62.5mM Tris -HCl (pH 6.8), 0.002% Bromophenol blue.DNase: None detectedRNase: None detectedProteases: None detectedStability (4°C) >1 Year
Catalog No. 50-196-778 Supplier Bioworld Supplier No. 105700181
Laemmli (SDS-Sample) 4X Buffer, Non-Reducing An electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Composition of this buffer is similar to the reducing buffer minus mercaptoethanol. Technical Information:Ultra Pure GradeContents of a 1X Solution: 2% SDS and 10% glycerol, 62.5mM Tris -HCl (pH 6.8), 0.002% Bromophenol blue.DNase: None detectedRNase: None detectedProteases: None detectedStability (4°C) >1 Year