Learn More
Boston Bioproducts Inc Laemmli SDS-Sample Buffer (6X, Reducing) - 25ml
Catalog No. NC9566545
Encompass
The 6X laemmli sample buffer is a critical component in the preparation of protein samples for SDS-PAGE, ensuring the resolution of protein bands. Its composition typically includes Tris-HCl, glycerol, SDS, and a tracking dye like Bromophenol Blue. In the reducing Laemmli sample buffer, a reducing agent such as β-mercaptoethanol or dithiothreitol (DTT) is added to break disulfide bonds within protein subunits, allowing them to separate by size in electrophoresis.
Catalog No. NC9566545 Supplier Boston Bioproducts Inc Supplier No. BP111R25ML
The 6X laemmli sample buffer is a critical component in the preparation of protein samples for SDS-PAGE, ensuring the resolution of protein bands. Its composition typically includes Tris-HCl, glycerol, SDS, and a tracking dye like Bromophenol Blue. In the reducing Laemmli sample buffer, a reducing agent such as β-mercaptoethanol or dithiothreitol (DTT) is added to break disulfide bonds within protein subunits, allowing them to separate by size in electrophoresis.