Learn More
Medchemexpress LLC Laetanine | 72361-67-2 | 313.35 g·mol⁻¹ | C18H19NO4 | 10 MG
SDP
Catalog No. 5000289833
Encompass_Preferred
Laetanine is a noraporphine alkaloid isolated from Litsea laeta and reported in extracts of Phoebe tavoyana. It exhibits antiplasmodial activity and is supplied as a research reagent for biological evaluation.
  • Natural noraporphine alkaloid from plant sources.
  • Exhibits antiplasmodial activity reported in literature.
  • Chemical formula C18H19NO4 and molecular weight 313.35 g·mol⁻¹.
  • Provided in both solid milligram quantities and solution form for assay preparation.
Catalog No. 50-002-89833 Supplier Medchemexpress LLC Supplier No. HYN430710MG
Laetanine is a noraporphine alkaloid isolated from Litsea laeta and reported in extracts of Phoebe tavoyana. It exhibits antiplasmodial activity and is supplied as a research reagent for biological evaluation.
  • Natural noraporphine alkaloid from plant sources.
  • Exhibits antiplasmodial activity reported in literature.
  • Chemical formula C18H19NO4 and molecular weight 313.35 g·mol⁻¹.
  • Provided in both solid milligram quantities and solution form for assay preparation.