Learn More
Medchemexpress LLC Laetanine | 72361-67-2 | 5 MG
SDP
Catalog No. 5000418138
Encompass_Preferred
Laetanine is a noraporphine alkaloid derived from *Litsea laeta* and exhibits antiplasmodial activity. It is also found in the extract of the leaves of *Phoebe tavoyana (Meissn) Hook*. This compound has a molecular weight of 313.35, a chemical formula of C18H19NO4, and appears as a solid with a light brown to brown color.
  • Exhibits antiplasmodial activity
  • Noraporphine alkaloid
  • Derived from *Litsea laeta*
  • Also found in *Phoebe tavoyana (Meissn) Hook*
  • Classified as an alkaloid, isoquinoline alkaloid, phenol, and polyphenol
  • Initial source from plants: Lauraceae, *Lindera aggregata (Sims) Kosterm*
  • For research use only
Catalog No. 50-004-18138 Supplier Medchemexpress LLC Supplier No. HYN43075MG
Laetanine is a noraporphine alkaloid derived from *Litsea laeta* and exhibits antiplasmodial activity. It is also found in the extract of the leaves of *Phoebe tavoyana (Meissn) Hook*. This compound has a molecular weight of 313.35, a chemical formula of C18H19NO4, and appears as a solid with a light brown to brown color.
  • Exhibits antiplasmodial activity
  • Noraporphine alkaloid
  • Derived from *Litsea laeta*
  • Also found in *Phoebe tavoyana (Meissn) Hook*
  • Classified as an alkaloid, isoquinoline alkaloid, phenol, and polyphenol
  • Initial source from plants: Lauraceae, *Lindera aggregata (Sims) Kosterm*
  • For research use only