Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lafadofensine D-(-)-mandelic acid | 00-00-0 | 100.0% | C32H32F2N2O6 | 10MG
SDP

Catalog No. 5000260851
Encompass_Preferred

Lafadofensine (D-(-)-mandelic acid) is a research compound described as a monoamines reuptake inhibitor with reported activity after short-term administration. It is supplied as a high-purity material for laboratory research and includes recommended storage conditions for powder and solutions.

  • Reported purity 99.95%.
  • Molecular formula C32H32F2N2O6.
  • Molecular weight 578.60 g/mol.
  • Available in small milligram sizes (5 mg-100 mg); larger sizes available by quote.
  • Storage: powder at -20 °C for up to 3 years; in solvent at -80 °C for up to 6 months.

Catalog No. 50-002-60851 Supplier Medchemexpress LLC Supplier No. HY145577A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lafadofensine (D-(-)-mandelic acid) is a research compound described as a monoamines reuptake inhibitor with reported activity after short-term administration. It is supplied as a high-purity material for laboratory research and includes recommended storage conditions for powder and solutions.

  • Reported purity 99.95%.
  • Molecular formula C32H32F2N2O6.
  • Molecular weight 578.60 g/mol.
  • Available in small milligram sizes (5 mg-100 mg); larger sizes available by quote.
  • Storage: powder at -20 °C for up to 3 years; in solvent at -80 °C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.