Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lafadofensine (D-(-)-Mandelic acid) | >99.95% | C32H32F2N2O6 | 1 ML
SDP

Catalog No. 5000355868
Encompass_Preferred

Lafadofensine (D-(-)-Mandelic acid) is a monoamines reuptake inhibitor that has sufficient effects after short-term administration. It is supplied as a solid, white to off-white powder with a molecular weight of 578.60 and a chemical formula of C32H32F2N2O6. This product is for research use only and not intended for sale to patients. It may be a controlled substance and not for sale in certain territories.

  • Monoamines reuptake inhibitor
  • Sufficient effects after short-term administration
  • Soluble in DMSO at 250 mg/mL (432.08 mM)
  • Purity of >99.95%
  • Research use only
  • Shipping at room temperature (continental US)

Catalog No. 50-003-55868 Supplier Medchemexpress LLC Supplier No. HY145577A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lafadofensine (D-(-)-Mandelic acid) is a monoamines reuptake inhibitor that has sufficient effects after short-term administration. It is supplied as a solid, white to off-white powder with a molecular weight of 578.60 and a chemical formula of C32H32F2N2O6. This product is for research use only and not intended for sale to patients. It may be a controlled substance and not for sale in certain territories.

  • Monoamines reuptake inhibitor
  • Sufficient effects after short-term administration
  • Soluble in DMSO at 250 mg/mL (432.08 mM)
  • Purity of >99.95%
  • Research use only
  • Shipping at room temperature (continental US)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.