Learn More
Sterlitech Corporation Lafil 200-BioDolphin Large Capacity Suction, Vacuum Filtration System, AC110V/60HZ
SDP
Catalog No. 501952302
Encompass_Preferred
The BioDolphin suction system includes the Lafil 200 unit, BioDophin suction kit, and 4L PP Waste Bottle with Quick-Coupling Connector. Media aspiration is optimized by integrated vacuum source with waste bottle and suction pipette. The system is sold with a 4 L polypropylene waste bottle, with a 3.5 L glass bottle optional. Features include built-in overflow protection to prevent solution from flowing into the pump, an ergonomic handle to enable continuous suction with no button-pressing fatigue, and easy tip ejection. A built-in liquid collection tray catches liquid that may drip from the fittings. The waste bottle and all supplied fitting parts are autoclavable.
Catalog No. 50-195-2302 Supplier Sterlitech Corporation Supplier No. 19720301
The BioDolphin suction system includes the Lafil 200 unit, BioDophin suction kit, and 4L PP Waste Bottle with Quick-Coupling Connector. Media aspiration is optimized by integrated vacuum source with waste bottle and suction pipette. The system is sold with a 4 L polypropylene waste bottle, with a 3.5 L glass bottle optional. Features include built-in overflow protection to prevent solution from flowing into the pump, an ergonomic handle to enable continuous suction with no button-pressing fatigue, and easy tip ejection. A built-in liquid collection tray catches liquid that may drip from the fittings. The waste bottle and all supplied fitting parts are autoclavable.