Learn More
Sterlitech Corporation Lafil 300C, PTFE Coated Chemical Resistant Vacuum Pump, AC110V, 60Hz
SDP
Catalog No. 501952304
Encompass_Preferred
Application:Ideal for the purification of organic, corrosive liquid such as solvent (mobile phase) for HPLC analysis.
Catalog No. 50-195-2304 Supplier Sterlitech Corporation Supplier No. 19733011
Application:Ideal for the purification of organic, corrosive liquid such as solvent (mobile phase) for HPLC analysis.