Learn More
Sterlitech Corporation Lafil 300C-VF12, PTFE Coated Chemical Resistant Vacuum Filtration System, AC110V, 60Hz
SDP
Catalog No. 501952299
Encompass_Preferred
Consits of:* Lafil 300C vacuum pump* VF12 glass filter holder, adaptor and 1 liter bottle Application:Ideal for the purification of organic, corrosive liquid such as solvent (mobile phase) for HPLC analysis.
Catalog No. 50-195-2299 Supplier Sterlitech Corporation Supplier No. 19733111
Consits of:* Lafil 300C vacuum pump* VF12 glass filter holder, adaptor and 1 liter bottle Application:Ideal for the purification of organic, corrosive liquid such as solvent (mobile phase) for HPLC analysis.