Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Laflunimus | 147076-36-6 | MFCD28502173 | 99.7% | 310.27 g/mol | C15H13F3N2O2 | 10 MG
SDP

Catalog No. 5000248843
Encompass_Preferred

Laflunimus is an orally active immunosuppressive small molecule that inhibits dihydroorotate dehydrogenase (DHODH) and prostaglandin endoperoxide H synthases (PGHS-1 and PGHS-2). It suppresses immunoglobulin secretion (reported IC50 ≈ 2-2.5 μM) and is used in research probing DHODH-mediated immunomodulation and related biochemical pathways.

  • High purity: 99.7%.
  • Molecular formula C15H13F3N2O2; molecular weight 310.27 g/mol.
  • Targets DHODH and PGHS-1/2, suitable for mechanism-of-action studies.
  • Appearance: solid, yellow to khaki.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years).
  • Suitable for biochemical and cell-based assays.

Catalog No. 50-002-48843 Supplier Medchemexpress LLC Supplier No. HY10181310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laflunimus is an orally active immunosuppressive small molecule that inhibits dihydroorotate dehydrogenase (DHODH) and prostaglandin endoperoxide H synthases (PGHS-1 and PGHS-2). It suppresses immunoglobulin secretion (reported IC50 ≈ 2-2.5 μM) and is used in research probing DHODH-mediated immunomodulation and related biochemical pathways.

  • High purity: 99.7%.
  • Molecular formula C15H13F3N2O2; molecular weight 310.27 g/mol.
  • Targets DHODH and PGHS-1/2, suitable for mechanism-of-action studies.
  • Appearance: solid, yellow to khaki.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years).
  • Suitable for biochemical and cell-based assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.