Learn More
Medchemexpress LLC Laflunimus | 147076-36-6 | MFCD28502173 | 99.7% | 310.27 g/mol | C15H13F3N2O2 | 50 MG
SDP
Catalog No. 5000248845
Encompass_Preferred
Laflunimus (HR325) is an orally active inhibitor of dihydroorotate dehydrogenase (DHODH) used as a research reagent to study immunomodulation and nucleotide biosynthesis. It is supplied as a solid research compound with reported high purity and supporting analytical data for use in biochemical and cellular assays.
  • Orally active DHODH inhibitor for immunomodulatory studies.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple solid pack sizes, including 50 mg.
  • Characterized by CAS 147076-36-6 and molecular formula C15H13F3N2O2.
  • Supplied with supporting analytical documentation, such as RP-HPLC.
Catalog No. 50-002-48845 Supplier Medchemexpress LLC Supplier No. HY10181350MG
Laflunimus (HR325) is an orally active inhibitor of dihydroorotate dehydrogenase (DHODH) used as a research reagent to study immunomodulation and nucleotide biosynthesis. It is supplied as a solid research compound with reported high purity and supporting analytical data for use in biochemical and cellular assays.
  • Orally active DHODH inhibitor for immunomodulatory studies.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple solid pack sizes, including 50 mg.
  • Characterized by CAS 147076-36-6 and molecular formula C15H13F3N2O2.
  • Supplied with supporting analytical documentation, such as RP-HPLC.