Learn More
Apexbio Technology LLC Lafutidine 118288-08-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B15755.1
Encompass_Preferred
Lafutidine (CAS 118288-08-7) is a histamine H2 receptor antagonist that functions by inhibiting histamine-induced gastric acid secretion Through selective blockade of H2 receptors on gastric parietal cells it reduces basal and stimulated levels of gastric acid production Lafutidine is utilized in biomedical research exploring gastric physiology receptor signaling pathways and gastrointestinal pharmacology particularly in contexts related to acid-related disorders
Catalog No. 50-000-62465