Learn More
Enzo Life Sciences LAG-3 (human) monoclonal antibody (11E3) (preservative free) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804805PFC100
Encompass_Preferred
Clone: 11E3. Isotype: IgG1. Immunogen: Recombinant human LAG-3 (lymphocyte activation gene-3). Source: Purified from the hybridoma culture supernatant by Protein G Fast Flow Column. Species reactivity: Human, Monkey. Specificity: Recognizes the first N-terminal D1 domain of LAG-3. Applications: ELISA, ICC, IHC (FS), IP, WB, FUNC. Formulation: Liquid. In PBS containing 10% glycerol.
Catalog No. 50-200-9976