Learn More
Enzo Life Sciences LAG-3 (human) monoclonal antibody (17B4) (biotin conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804806BC100
Encompass_Preferred
Clone: 17B4. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to 30 aa (GPPAAAPGHPLAPGPHPAAPSSWGPRPRRY) from the first N-terminal D1 domain of human LAG-3 (lymphocyte activation gene-3). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: Recognizes the 30 aa extra-loop of the first N-terminal D1 domain of LAG-3. Applications: ELISA, WB. Purity: Purified from concentrated hybridoma tissue culture supernatant by Protein A Sepharose™ CL-4B Flow Fast Column. Formulation: Liquid. In PBS containing 50mM Tris-HCl, 1% BSA and 0.02% sodium azide.
Catalog No. 50-200-9977