Learn More
AdipoGen LAG-3 h mAb 17B4blocking
SDP
Supplier:  AdipoGen AG20B0012PFC500
Encompass_Preferred
Monoclonal Antibody. Recognizes human LAG-3. Isotype Mouse IgG1. Clone 17B4. Applications FACS, FUNC Blocking, ICC, IHC, IP, WB. Liquid. In PBS. Lymphocyte activation gene 3 LAG-3 CD223 plays an important role in negatively regulating T cell proliferation, function and homeostasis. It is required for maximal natural and induced regulatory T cell Treg function. LAG-3 is closely related to the T cell co-receptor CD4 and binds to MHC class II molecules but with a significantly higher affinity than CD4.
Catalog No. 50-256-3383