Learn More
Enzo Life Sciences LAG-3 (human) recombinant monoclonal antibody (L4-PL33) (ATTO647N conjugate) (100 µg)
SDP
Supplier:  Enzo Life Sciences ENZABS677TS0100
Encompass_Preferred
Synonyms: Lymphocyte activation gene-3, FDC protein, CD223. Clone: 17B4. Host: Mouse. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to 30 aa (GPPAAAPGHPLAPGPHPAAPSSWGPRPRRY) from the first N-terminal D1 domain of human LAG-3 (lymphocyte activation gene-3). UniProt ID: P18627. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Applications: Flow Cytometry, ICC. Purification: Protein A affinity purified. Formulation: Liquid. In PBS containing 50mM Tris, 1% BSA and 0.02% sodium azide.
Catalog No. 50-210-0440