Learn More
Enzo Life Sciences LAG-3 (human) recombinant monoclonal antibody (L4-PL33) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6770100
Encompass_Preferred
Clone: L4-PL33. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to 30 aa (GPPAAAPGHPLAPGPHPAAPSSWGPRPRRY) from the first N-terminal D1 domain of human LAG-3 (lymphocyte activation gene-3). Species reactivity: Human. Specificity: Recognizes the 30 aa extra-loop of the first N-terminal D1 domain of LAG-3. Applications: ELISA, Flow Cytometry, ICC, IHC (FS), IP, WB. Purity: Purified. Formulation: Liquid. In PBS containing 10% glycerol and 0.02% sodium azide.
Catalog No. 50-201-3205