Learn More
AdipoGen anti-LAG-3 (human), mAb (17B4) (ATTO 488) (100 tests)
SDP
Supplier:  AdipoGen AG20B0012TDT100
Encompass_Preferred
Monoclonal Antibody. Recognizes human LAG-3. Isotype: Mouse IgG1. Clone: 17B4. Applications: FACS, ICC. Liquid. In PBS containing 0.02% sodium azide. Lymphocyte activation gene 3 (LAG-3; CD223) plays an important role in negatively regulating T cell proliferation, function and homeostasis. It is required for maximal natural and induced regulatory T cell (Treg) function. LAG-3 is closely related to the T cell co-receptor CD4 and binds to MHC class II molecules but with a significantly higher affinity than CD4.
Catalog No. 50-205-2907